missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium nitride, 99.4% (metals basis), Thermo Scientific Chemicals

$137.88 - $520.75

Chemical Identifiers

CAS 26134-62-3
Molecular Formula Li3N
Molecular Weight (g/mol) 34.83
MDL Number MFCD00016186
InChI Key IDBFBDSKYCUNPW-UHFFFAOYSA-N
Synonym lithium nitride, trilithium nitride, li3n, lithium nitride li3n, nitrure de lithium, li3-n, nitruro de litio spanish, nitrure de lithium french, lithium nitride,anh, nli3
PubChem CID 520242
SMILES [Li]N([Li])[Li]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3547909
View Documents
Thermo Scientific Chemicals
03547909
10 g
Each for $137.88
 
AA3547918
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
03547918
50 g
Each for $520.75
 
Description

Description

Lithium nitride is used in metallurgy and chemical synthesis. It is also used to store hydrogen and acts as a source of the nitride ion. It is involved in the preparation of lithium hydride and lithium amide as well as a reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

26134-62-3
34.83
IDBFBDSKYCUNPW-UHFFFAOYSA-N
520242
Li3N
MFCD00016186
lithium nitride, trilithium nitride, li3n, lithium nitride li3n, nitrure de lithium, li3-n, nitruro de litio spanish, nitrure de lithium french, lithium nitride,anh, nli3
[Li]N([Li])[Li]

Specifications

845°C
Powder
99.4% (metals basis)
MFCD00016186
Insoluble in organic solvents.
[Li]N([Li])[Li]
34.83
34.82
1.30 g/mL
26134-62-3
10 g
Li3N
lithium nitride, trilithium nitride, li3n, lithium nitride li3n, nitrure de lithium, li3-n, nitruro de litio spanish, nitrure de lithium french, lithium nitride,anh, nli3
IDBFBDSKYCUNPW-UHFFFAOYSA-N
Lithium nitride
520242
Moisture sensitive
Lithium nitride
Safety and Handling

Safety and Handling

P210-P222-P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H250-H260-H314-H335

DOTInformation : Hazard Class: 4.3; Packaging Group: I

EINECSNumber : 247-475-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only