Learn More
Lithium oxalate, 99+%, Thermo Scientific Chemicals
$79.99 - $646.47
Chemical Identifiers
CAS | 553-91-3 |
---|---|
Molecular Formula | C2Li2O4 |
Molecular Weight (g/mol) | 101.90 |
MDL Number | MFCD00040596 |
InChI Key | YNQRWVCLAIUHHI-UHFFFAOYSA-L |
Synonym | lithium oxalate, dilithium oxalate, ethanedioic acid, dilithium salt, unii-k737ot0e73, oxalic acid dilithium salt, ethanedioic acid, lithium salt 1:2, oxalic acid dilithium, acmc-1ctwf, c2o4.2li, ksc492a1t |
PubChem CID | 68383 |
SMILES | [Li+].[Li+].[O-]C(=O)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1342618
|
Thermo Scientific Chemicals
01342618 |
50 g |
Each for $79.99
|
|
|||||
AA1342630
|
Thermo Scientific Chemicals
01342630 |
250 g |
Each for $226.45
|
|
|||||
AA13426A1
|
Thermo Scientific Chemicals
013426A1 |
1 kg |
Each for $646.47
|
|
|||||
Description
Lithium oxalate is used as lithium battery electrolyte. It is also used as an analytical reagent and reducing reagent. Further, it is used as an intermediate in the manufacture of other chemicals such as pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
553-91-3 | |
101.90 | |
YNQRWVCLAIUHHI-UHFFFAOYSA-L | |
68383 |
C2Li2O4 | |
MFCD00040596 | |
lithium oxalate, dilithium oxalate, ethanedioic acid, dilithium salt, unii-k737ot0e73, oxalic acid dilithium salt, ethanedioic acid, lithium salt 1:2, oxalic acid dilithium, acmc-1ctwf, c2o4.2li, ksc492a1t | |
[Li+].[Li+].[O-]C(=O)C([O-])=O |
Specifications
553-91-3 | |
2.12 g/mL | |
MFCD00040596 | |
UN3288 | |
lithium oxalate, dilithium oxalate, ethanedioic acid, dilithium salt, unii-k737ot0e73, oxalic acid dilithium salt, ethanedioic acid, lithium salt 1:2, oxalic acid dilithium, acmc-1ctwf, c2o4.2li, ksc492a1t | |
YNQRWVCLAIUHHI-UHFFFAOYSA-L | |
dilithium;oxalate | |
68383 | |
≥99% | |
Lithium oxalate |
(decomposition) | |
C2Li2O4 | |
50 g | |
14,5537 | |
Soluble in water. | |
[Li+].[Li+].[O-]C(=O)C([O-])=O | |
101.90 | |
101.96 | |
Powder |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber : 209-054-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only