Learn More
Lithium perchlorate, 99+%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 194715000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Oxidation reagent for analysisSpecifications
Lithium perchlorate | |
p.a. | |
Colorless to Yellow | |
500 g | |
ClLiO4 | |
MFCD00011079 | |
15, 5595 | |
Solubility in water: 600g/L (25°C). Other solubilities: 1520g/L ethanol (25°C), 1820g/L methanol (25°C), 1140g/L ether (25°C), soluble in acetone and ethyl acetate | |
[Li+].[O-]Cl(=O)(=O)=O | |
106.39 | |
106.39 | |
Analytical |
236.0°C | |
7791-03-9 | |
Liquid | |
99+% | |
LiClO4 | |
17,167 | |
lithium perchlorate, perchloric acid, lithium salt, lithiumperchlorate, unii-q86se98c9c, lithium 1+ ion perchlorate, lithium perchlorate, anhydrous, lithium cloricum, lithium per chlorate, liclo4, perchloric acid, lithium salt 1:1 | |
JAWYRNYHJJDXHX-UHFFFAOYSA-M | |
lithium;perchlorate | |
23665649 | |
≥99% | |
Plastic bottle |
Chemical Identifiers
7791-03-9 | |
106.39 | |
JAWYRNYHJJDXHX-UHFFFAOYSA-M | |
23665649 | |
[Li+].[O-]Cl(=O)(=O)=O |
ClLiO4 | |
MFCD00011079 | |
lithium perchlorate, perchloric acid, lithium salt, lithiumperchlorate, unii-q86se98c9c, lithium 1+ ion perchlorate, lithium perchlorate, anhydrous, lithium cloricum, lithium per chlorate, liclo4, perchloric acid, lithium salt 1:1 | |
lithium;perchlorate |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause fire or explosion; strong oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/ combustible materials.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 232-237-2
TSCA : TSCA
RUO – Research Use Only