Learn More
Lithium periodate dihydrate, Reagent Grade
CAS: 307310-74-3 | ILiO4 | 197.84 g/mol
Supplier: Thermo Scientific Chemicals 03935518
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Lithium periodate dihydrate | |
| 307310-74-3 | |
| 50 g | |
| MFCD00049474 | |
| lithium periodate, lithotab io4, acmc-20akme, lithium 1+ ion periodate, lithium 1+ periodate, lithium periodate lih4io6 | |
| SYWXNZXEJFSLEU-UHFFFAOYSA-M | |
| lithium;periodate | |
| 23711816 | |
| Reagent |
| >370°C | |
| Powder | |
| ILiO4 | |
| UN1479 | |
| Soluble in water. | |
| [Li+].[O-]I(=O)(=O)=O | |
| 197.84 | |
| 233.87 (197.84 Anhydrous) | |
| Odorless |
Chemical Identifiers
| 307310-74-3 | |
| 197.84 | |
| SYWXNZXEJFSLEU-UHFFFAOYSA-M | |
| 23711816 | |
| [Li+].[O-]I(=O)(=O)=O |
| ILiO4 | |
| MFCD00049474 | |
| lithium periodate, lithotab io4, acmc-20akme, lithium 1+ ion periodate, lithium 1+ periodate, lithium periodate lih4io6 | |
| lithium;periodate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only