missing translation for 'onlineSavingsMsg'
Learn More

Lithium periodate dihydrate, Reagent Grade

CAS: 307310-74-3 | ILiO4 | 197.84 g/mol

Supplier:  Thermo Scientific Chemicals 03935530

 View more versions of this product

Catalog No. AA3935530


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lithium periodate dihydrate
307310-74-3
250 g
MFCD00049474
lithium periodate, lithotab io4, acmc-20akme, lithium 1+ ion periodate, lithium 1+ periodate, lithium periodate lih4io6
SYWXNZXEJFSLEU-UHFFFAOYSA-M
lithium;periodate
23711816
Reagent
>370°C
Powder
ILiO4
UN1479
Soluble in water.
[Li+].[O-]I(=O)(=O)=O
197.84
233.87 (197.84 Anhydrous)
Odorless

Chemical Identifiers

307310-74-3
197.84
SYWXNZXEJFSLEU-UHFFFAOYSA-M
23711816
[Li+].[O-]I(=O)(=O)=O
ILiO4
MFCD00049474
lithium periodate, lithotab io4, acmc-20akme, lithium 1+ ion periodate, lithium 1+ periodate, lithium periodate lih4io6
lithium;periodate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only