missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium phosphate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01074322

 View more versions of this product

Catalog No. AA1074322


Explore available promotions

Description

Description

Lithium phosphate batteries batteries are now widely used as a staple for cell phones, laptops and some other gadgets. Used in low-expansion porcelain enamel glazes and polymer intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lithium phosphate
10377-52-3
Powder
99.99% (Metals basis)
Li3O4P
lithium phosphate, lithium orthophosphate, trilithium orthophosphate, trilithium phosphate, phosphoric acid, trilithium salt, unii-2qm4k05q74, lithiumphosphate, trilithotab phosphate, phosphoric acid, lithium salt 1:3, li3po4
TWQULNDIKKJZPH-UHFFFAOYSA-K
trilithium;phosphate
165867
99.99%
Odorless
837°C
White
100 g
(metals basis)
MFCD00016187
Poor solubility in water. Insoluble in alcohols.
[Li+].[Li+].[Li+].[O-]P([O-])([O-])=O
115.79
115.79
Puratronic™
2.54

Chemical Identifiers

10377-52-3
115.79
TWQULNDIKKJZPH-UHFFFAOYSA-K
165867
Li3O4P
MFCD00016187
lithium phosphate, lithium orthophosphate, trilithium orthophosphate, trilithium phosphate, phosphoric acid, trilithium salt, unii-2qm4k05q74, lithiumphosphate, trilithotab phosphate, phosphoric acid, lithium salt 1:3, li3po4
[Li+].[Li+].[Li+].[O-]P([O-])([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 233-823-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only