Learn More
Lithium sulfate, 99%, extra pure, anhydrous
CAS: 10377-48-7 | Li2O4S | 109.94 g/mol
$178.92 - $1994.98
Chemical Identifiers
| CAS | 10377-48-7 |
|---|---|
| Molecular Formula | Li2O4S |
| Molecular Weight (g/mol) | 109.94 |
| MDL Number | MFCD00011086 |
| InChI Key | INHCSSUBVCNVSK-UHFFFAOYSA-L |
| Synonym | lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous |
| PubChem CID | 66320 |
| ChEBI | CHEBI:53474 |
| SMILES | [Li+].[Li+].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC218322500
|
Thermo Scientific Chemicals
218322500 |
250 g | Plastic bottle |
Each for $178.92
|
|
||||
|
AC218320010
|
Thermo Scientific Chemicals
218320010 |
1 kg | Plastic bottle |
Each for $455.72
|
|
||||
|
AC218320051
|
Thermo Scientific Chemicals
218320051 |
5 kg | Plastic Drum |
Each for $1,994.98
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10377-48-7 | |
| 109.94 | |
| INHCSSUBVCNVSK-UHFFFAOYSA-L | |
| 66320 | |
| [Li+].[Li+].[O-]S([O-])(=O)=O |
| Li2O4S | |
| MFCD00011086 | |
| lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous | |
| CHEBI:53474 |
Specifications
| 859.0°C | |
| White | |
| 250 g | |
| Li2O4S | |
| MFCD00011086 | |
| lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous | |
| INHCSSUBVCNVSK-UHFFFAOYSA-L | |
| dilithium;sulfate | |
| 66320 | |
| 109.94 | |
| Extra Pure | |
| 2.22 | |
| Lithium sulfate, 99% |
| 10377-48-7 | |
| Crystalline Powder | |
| 99% | |
| Li2SO4 | |
| 15,5597 | |
| Solubility in water: 25.7 g/100mL (25°C). Other solubilities: practically insoluble in alcohol | |
| [Li+].[Li+].[O-]S([O-])(=O)=O | |
| 109.94 | |
| CHEBI:53474 | |
| 99% | |
| Plastic bottle | |
| 2.2200g/mL |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr
GHS Signal Word: Warning
EINECSNumber : 233-820-4
RTECSNumber : OJ6419000
TSCA : TSCA
RUO – Research Use Only