missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium sulfate monohydrate, 99+%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 218331000

 View more versions of this product

Catalog No. AC218331000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

Lithium sulfate monohydrate
10102-25-7
Crystalline Powder
99+%
LI2SO4·H2O
15, 5597
Solubility in water: 295.5g/L (20°C)
[Li+].[Li+].O.[O-]S([O-])(=O)=O
127.95
127.96
Pure
Plastic bottle
860.0°C
White
100 g
H2Li2O5S
MFCD00149766
lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
RKGLUDFWIKNKMX-UHFFFAOYSA-L
dilithium;sulfate;hydrate
165815
≥99%
13.5% to 15% (600°C, 2 hrs)

Chemical Identifiers

10102-25-7
127.95
RKGLUDFWIKNKMX-UHFFFAOYSA-L
165815
[Li+].[Li+].O.[O-]S([O-])(=O)=O
H2Li2O5S
MFCD00149766
lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
dilithium;sulfate;hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only