Learn More
Lithium sulfide, 99.9% (metals basis)
CAS: 12136-58-2 | Li2S | 45.94 g/mol
$104.15 - $3571.22
Chemical Identifiers
| CAS | 12136-58-2 |
|---|---|
| Molecular Formula | Li2S |
| Molecular Weight (g/mol) | 45.94 |
| MDL Number | MFCD00011085 |
| InChI Key | GLNWILHOFOBOFD-UHFFFAOYSA-N |
| Synonym | lithium sulfide, lithium sulfide li2s, lithium sulphide, anhydrous, li2s, zwdbutfcwlvlcq-uhfffaoysa-n, lithium sulfide trace metals basis |
| PubChem CID | 64734 |
| SMILES | [Li+].[Li+].[S--] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1283904
|
Thermo Scientific Chemicals
01283904 |
2 g |
Each for $104.15
|
|
|||||
|
AA1283909
|
Thermo Scientific Chemicals
01283909 |
10 g |
Each for $321.29
|
|
|||||
|
AA1283918
|
Thermo Scientific Chemicals
01283918 |
50 g |
Each for $1,025.64
|
|
|||||
|
AA1283930
|
Thermo Scientific Chemicals
01283930 |
250 g |
Each for $3,571.22
|
|
|||||
Description
It act as a superconductor and widely used in lithium-sulfur batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12136-58-2 | |
| 45.94 | |
| GLNWILHOFOBOFD-UHFFFAOYSA-N | |
| 64734 |
| Li2S | |
| MFCD00011085 | |
| lithium sulfide, lithium sulfide li2s, lithium sulphide, anhydrous, li2s, zwdbutfcwlvlcq-uhfffaoysa-n, lithium sulfide trace metals basis | |
| [Li+].[Li+].[S--] |
Specifications
| 900°C to 975°C | |
| Yellow | |
| 2 g | |
| Li2S | |
| lithium sulfide, lithium sulfide li2s, lithium sulphide, anhydrous, li2s, zwdbutfcwlvlcq-uhfffaoysa-n, lithium sulfide trace metals basis | |
| GLNWILHOFOBOFD-UHFFFAOYSA-N | |
| dilithium(1+) sulfanediide | |
| 64734 | |
| Like rotten eggs | |
| Yellow powder | |
| Lithium sulfide |
| 12136-58-2 | |
| Powder | |
| 99.9% (Metal basis) | |
| MFCD00011085 | |
| soluble in water and ethanol | |
| [Li+].[Li+].[S--] | |
| 45.94 | |
| 45.95 | |
| Moisture sensitive | |
| 1.66 g/cm3 at 20°C |
Safety and Handling
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314-H335
DOTInformation : DOT Class: 8; Packaging Group: II
EINECSNumber : 235-228-1
RTECSNumber : OJ6439500
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only