missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium tri-sec-butylborohydride, 1.0M solution in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

As a reducing agent of olefinic groups and ketones

Supplier:  Thermo Scientific Chemicals 043034AP

 View more versions of this product

Catalog No. AA43034AP


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications

Specifications

Lithium tri-sec-butylborohydride
Colorless to Yellow
−17°C (1°F)
C12H28BLi
MFCD00011708
UN3394
Moisture sensitive
lithium tri-sec-butylhydroborate, l-selectridesolution, lithium tri-sec-butylborohydride solution, l-selectride™ solution, lithium tri butan-2-yl boron 1-, lithium tri-sec-butylborohydride l-selectride , 1m in thf, lithium tri-sec-butylborohydride 1.0 m solution in thf, spcseal, lithium tri-sec-butylborohydride, 1.0m solution in thf, packaged under argon in resealable chemseal bottles
ACJKNTZKEFMEAK-UHFFFAOYNA-N
lithium(1+) tris(butan-2-yl)boranuide
190.11
Liquid
38721-52-7
0.870 g/mL
ChemSeal™ bottles
LiB[CH(CH3)CH2CH3]3H
500 mL
4007327
14,8427
Miscible with tetrahydrofuran.
[Li+].CCC(C)[BH-](C(C)CC)C(C)CC
190.11
1.0M solution in THF

Chemical Identifiers

38721-52-7
190.11
ACJKNTZKEFMEAK-UHFFFAOYNA-N
lithium(1+) tris(butan-2-yl)boranuide
C12H28BLi
MFCD00011708
lithium tri-sec-butylhydroborate, l-selectridesolution, lithium tri-sec-butylborohydride solution, l-selectride™ solution, lithium tri butan-2-yl boron 1-, lithium tri-sec-butylborohydride l-selectride , 1m in thf, lithium tri-sec-butylborohydride 1.0 m solution in thf, spcseal, lithium tri-sec-butylborohydride, 1.0m solution in thf, packaged under argon in resealable chemseal bottles
[Li+].CCC(C)[BH-](C(C)CC)C(C)CC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H250-H260-H314-H318
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c

H225-H260-H314-H335-H336-H351

DOTInformation : Hazard Class: 4.3; Packaging Group: I

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only