Learn More
Lithium (trimethylsilyl)acetylide, 0.5M solution in THF, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 380658000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Lithium (trimethylsilyl)acetylide | |
800mL | |
MFCD00075059 | |
Solubility in water: reacts | |
[Li+].C[Si](C)(C)C#[C-] | |
104.15 | |
104.15 | |
0.88 |
54655-07-1,109-99-9 | |
C5H9LiSi | |
trimethylsilyl ethynyllithium, lithium, trimethylsilyl ethynyl, lithium 1+ ion trimethylsilyl ethyne, lithium trimethylsilylacetylide, lithio trimethylsilyl acetylene, lithium trimethylsilyl acetylide, lithium ethynyl trimethyl silane, lithium trimethylsilylacetylenide, zvxxeonxfwsciz-uhfffaoysa-n | |
WDDOQHLJFOUQMW-UHFFFAOYSA-N | |
lithium;ethynyl(trimethyl)silane | |
3431600 | |
AcroSeal™ Glass Bottle | |
0.8800g/mL |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
May form explosive peroxides.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
GHS Signal Word: Danger
RUO â Research Use Only