Learn More
Lithium wire, 3.2mm (0.125in) dia, 99.8%, Na 0.03% max, Thermo Scientific Chemicals
$821.78 - $2465.34
Chemical Identifiers
CAS | 7439-93-2 |
---|---|
Molecular Formula | Li |
Molecular Weight (g/mol) | 6.94 |
MDL Number | MFCD00134051 |
InChI Key | WHXSMMKQMYFTQS-UHFFFAOYSA-N |
Synonym | litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549 |
PubChem CID | 3028194 |
ChEBI | CHEBI:30145 |
IUPAC Name | lithium |
SMILES | [Li] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-93-2 | |
6.94 | |
WHXSMMKQMYFTQS-UHFFFAOYSA-N | |
3028194 | |
lithium |
Li | |
MFCD00134051 | |
litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549 | |
CHEBI:30145 | |
[Li] |
Specifications
7439-93-2 | |
99.8% | |
MFCD00134051 | |
litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549 | |
[Li] | |
6.94 | |
CHEBI:30145 | |
Annealed Wire |
0.53 | |
Li | |
25 g | |
WHXSMMKQMYFTQS-UHFFFAOYSA-N | |
lithium | |
3028194 | |
6.941 | |
Lithium, 3.2mm (0.125 in.) Dia. |
Safety and Handling
P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H260-H314
DOTInformation : Hazard Class: 4.3; Packaging Group: I
EINECSNumber : 231-102-5
RTECSNumber : OJ5540000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only