Thermo Scientific™ LN2 Supply Tanks
Transport cryogenic industrial gases with these LN2 Supply tanks, which are designed to be both durable and user-friendly.
$14264.10 - $24702.34
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
TF8127
|
Thermo Scientific™
8127 |
50 L |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
TF8121
|
Thermo Scientific™
8121 |
230 L |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Transport cryogenic industrial gases with Thermo Scientific™ LN2 Supply tanks, which are designed to be both durable and user-friendly.
For long-term storage of samples in cryogenic freezers, or controlling temperature during freezing procedures utilizing controlled-rate freezing equipment.- Rated at 22psi (151.7kPa)
- Evaporate 2% of volume per day
- Warranty varies by country, please contact us for details
Specifications
LN2 Supply Tank |