missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Alkalinity
Accurate test kits for routine water analysis
Supplier: Lovibond™ 147450
Includes:
- Aqua Comparator Housing
- Aqua Comparator Disc
- Alkacheck Tablets (100 pack)
- Sample Cells (2)
- Tablet Crusher
- Instructions
- Carrying Case
Description
Accurate test kits for routine water analysis
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
| ±5% | |
| Aqua comparator™ housing, aqua comparator™ disc, alkacheck tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case | |
| 20 to 240mg/L |
| Routine water analysis | |
| 100 | |
| Alkalinity Test Kit |