Lovibond™ CHECKIT™ Comparator Test Kits: Total Hardness
Accurate test kits for routine water analysis
Supplier:  Lovibond™ 414250
Catalog No. 09-053-135
Explore available promotions
Includes:
  • Calibrated sample container, 100mL
  • Total Hardness LR (BW) Tablets (200 tablets)
  • Instructions
  • Storage Case
Description
Description
Specifications
Specifications
Routine water analysis
100
LR Total Hardness Mini Kit
100mL calibrated sample container, total hardness LR tablets, instructions, storage case
1 to 50mg/L