Lovibond™ COD Tube Tests, HR
Premeasured unit dose reagent for water quality testing
Supplier:  Lovibond™ 2420722
Catalog No. 09-054-105
Explore available promotions
Description
Description
  • For use in SC400™ Colorimeters and Aqua Comparator™ Test Kits
  • Also suitable for use in Hach™ instruments and test kits
  • Easy-to-open, sealed vials eliminate risk of cross-contamination
  • Extended shelf life of up to five years
For Municipal Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted test method
Specifications
Specifications
US EPA
SC400 colorimeters and Aqua comparator™ test kits, Hach™ instruments
0 to 15000mg/L
Municipal wastewater testing, industrial wastewater and process control testing, environmental water testing
25
Dichromate/H2SO4