missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lovibond™ Colorimeter: COD
These rugged, waterproof, single parameter colorimeters are ideal for laboratory and field testing
Supplier: Lovibond™ 276120
Description
- Ranges 0-150mg/L, 0-1,500mg/L and 0-15,000mg/L
- Method uses Dichromate/H2SO4 reagent
- COD test tube reagents, 25 or 150 tests, sold separately
- Reactor required to heat (digest) sample at 150°C
- Large, backlit display; real time clock; data logging capabilities
- Platform accommodates one LED light source and one photo diode
- Power: 9V battery
- Platform can accommodate up to 3 LED light sources and photo diodes
- Power: Four AAA batteries
- Can export data using the IRiM
Specifications
3% Full scale | |
CE | |
4 x AAA | |
6.1 x 3 x 1.5 in. (15.5 x 7.6 x 3.8 cm) | |
LED | |
430, 610 nm | |
1.5 in. | |
Instrument, 16mm tube adapter, batteries, tube brush, instruction manual and a carrying case | |
15.5 cm | |
0.01 A | |
2 Years | |
250 g | |
7.6 cm |
Yes | |
Automatic; 16 data sets with time and date stamp | |
Backlit LCD | |
IP68 | |
COD Colorimeter | |
Laboratory and field testing | |
3.8 cm | |
6.1 in. | |
0 to 150 mg/L, 0 to 1500 mg/L, 0 to 15000 mg/L | |
5°C to 40°C | |
0.55 lb. | |
3 in. |