Lovibond™ Total Nitrogen Reagent Set, for Tube Testing, High Range
Premeasured unit dose reagent for water quality testing
Supplier: Lovibond™ 535560
Includes: Total N Persulfate Reagent, Total Nitrogen Reagent A, Total Nitrogen Reagent B, Total Nitrogen Hydroxide HR Tubes, Total Nitrogen Acid LR/HR Tubes, Deionized Water, 100mL
Description
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
Total N persulfate reagent (50 pack), total nitrogen reagent A (50 pack), total nitrogen reagent B (50 pack), 50 ct. total nitrogen hydroxide HR tubes, 50 ct. total nitrogen Acid LR/HR tubes, 100mL deionized water | |
0 to 150mg/L | |
High Range Total Nitrogen Reagent Set |
Hach™ and other manufacturers test kits, colorimeters and spectrophotometers, reactor | |
25 | |
Persulfate Digestion |