Corning™ PYREX™ Low Form Griffin Beakers
Offers optimum balance between thermal shock resistance and mechanical strength
Supplier:  Corning™ 100030
 View more versions of this product
Catalog No. 02-540F
Explore available promotions
Description
Description
With pouring spout and extra-large marking spot
Corning™ PYREX™ Low Form Griffin Beakers offer optimum balance between thermal shock resistance and mechanical strength.
With pouring spout and extra-large marking spot.
Beakers, with spout, manufactured with uniform wall thickness, offers optimum balance between thermal shock resistance and mechanical strength. For convenience, the 250 through 4000 mL beakers have a double graduated metric scale to indicate approximate content. All sizes have an extra large marking spot. The 10 mL size is not graduated.
Compliance
Designed to meet ASTM™ E-960 requirements
Specifications
Specifications
1.01 oz.
3.5 cm
Borosilicate Glass
5 to 25 mL
5.3 cm
10 mL
12 Pk.
Reusable
ASTM E960
Griffin
1.37 in.
30 mL
For Optimum Balance Between Thermal Shock Resistance and Mechanical Strength
2.08 in.
Spout
White Marking
Beaker
Yes
Dual White Metric Scale