MilliporeSigma™ Supelco™ LpDNPH S10x Cartridge, Bed wt. 350 mg
Designed for sampling in ambient, indoor, and industrial atmospheres carbonyls
Supplier:  MilliporeSigma™ Supelco™ 505293
Catalog No. 111010044
Explore available promotions
Description
Description
LpDNPH S10x Cartridge is an air sampling device designed for sampling in ambient, indoor, and industrial atmospheres carbonyls. Carbonyls are trapped on a high purity silica adsorbent coated with 2,4-dinitrophenylhydrazine, where they are converted to the hydrazone derivatives. The derivatives are eluted from the cartridge in acetonitrile and are analyzed by HPLC.
Specifications
Specifications
Bed wt. 350 mg
10/Pk.
1.5 mL