missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

STERIS LpH™ III se Phenolic Disinfectant

This Phenolic Disinfectant is the latest generation of low pH disinfectant and is specifically formulated for the routine disinfection of hard, non-porous surfaces.

$439.63 - $4561.00

Specifications

Product Type Phenolic Disinfectant
Form Concentrated Liquid
Sterility Non-sterile
Concentration 0.8% v/v (1:128)
View More Specs

Products 3
Catalog Number Mfr. No. Volume (English) Price Quantity  
Catalog Number Mfr. No. Volume (English) Price Quantity  
17-463-009
View Documents
STERIS
647008CA
1 gal.
Case of 4 Each for $482.36
 
19-415-153
View Documents
STERIS
647005CA
5 gal.
Each for $439.63
 
19-415-154
View Documents
STERIS
647001CA
55 gal.
Each for $4,561.00
 
Description

Description

This STERIS™ LpH™ III se Disinfectant is specifically formulated for the routine disinfection of hard, non-porous surfaces. At a use dilution of rate of 0.8% v/v (1:128), the LpH™ III Disinfectants kill a broad spectrum of common bacteria and fungi. The acidic LpH™ III Disinfectant formulation can be used alone or in rotation with the Vesphene™ III Disinfectants. These products have an extensive documentation package to meet your validation objectives and are supported by the STERIS™ Technical Services team for individualized application and validation assistance.

How LpH™ III se Disinfectant Works

STERIS'™ LpH™ III se Phenolic Disinfectant is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.

Why LpH™ III se Disinfectant?

  • Efficacy
    • Phenolic active ingredients provide broad spectrum antimicrobial efficacy.
  • Productivity
    • Formulation includes surfactants for surface cleaning while disinfecting.
  • Flexibility
    • Ideal for use in a contamination control rotation.
  • Reliability
    • Complete lot traceability.
  • Assurance
    • Each lot documented by Certificate of Analysis.
Specifications

Specifications

Phenolic Disinfectant
Concentrated Liquid
Non-sterile
0.8% v/v (1:128)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'