Cytiva Luer Connectors for Chromatography Columns
For use with the HiTrap column family of columns
Supplier: Cytiva 18100368
Requires : Cytiva HiTrap Columns, sold separately.
Description
- HiTrap column inlets have 1/16 in. female threads, while the outlets have 1/16 in. male threads
Specifications
Tubing connecto r | |
Polypropylene | |
Connect tubing to botto m of column |
2 Pack | |
Column Accesory |