Cytiva Luer Connectors for Chromatography Columns
For use with the HiTrap column family of columns
Supplier: Cytiva 18102762
Requires : Cytiva HiTrap Columns, sold separately.
Description
- HiTrap column inlets have 1/16 in. female threads, while the outlets have 1/16 in. male threads
Specifications
Union | |
Polypropylene | |
Connect a syringe to the botto m of a HiTrap column |
2 Pack | |
Column Accesory |