missing translation for 'onlineSavingsMsg'
Learn More

Lugol's Iodine, 5.15%, USP Strong Iodine Solution, Reagents

Supplier:  Labchem Inc C7217850500C

Catalog No. 361059174

Only null left
Explore available promotions

Description

Description

  • Lugol's Iodine
  • USP Test Solution Formulation
  • Appearance: Dark Brown Liquid
  • Specific Gravity: 1.111
  • Size: 500 mL
  • Brand: Reagents
Specifications

Specifications

Liquid or Solid
Blend
Laboratory
500 mL
100%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions