missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Luminol 98.0+%, TCI America™

Supplier:  TCI America A530125G

 View more versions of this product

Catalog No. A530125G

Explore available promotions

Description

Description

Specifications

Specifications

Luminol [Chemiluminescence Reagent]
320°C
MFCD00006890
luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide
C1=CC2=C(C(=C1)N)C(=O)NNC2=O
177.163
177.16
Crystalline Powder
521-31-3
C8H7N3O2
25 g
HWYHZTIRURJOHG-UHFFFAOYSA-N
5-amino-2,3-dihydrophthalazine-1,4-dione
10638
≥98.0% (HPLC,T)

Chemical Identifiers

521-31-3
177.163
HWYHZTIRURJOHG-UHFFFAOYSA-N
10638
C1=CC2=C(C(=C1)N)C(=O)NNC2=O
C8H7N3O2
MFCD00006890
luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide
5-amino-2,3-dihydrophthalazine-1,4-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : TH8890060

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions