Greiner Bio-One LUMITRAC™ 96-well Strip Plate, Single-break
For use in research. Greiner Bio-One LUMITRAC™ 96-Well Strip Plate, Single-break can be broken off separately and number of tests performed can be adjusted to number of samples.
Supplier:  Greiner Bio-One 705092
 View more versions of this product
Catalog No. 07000786
Explore available promotions
Description
Description
  • 1 x 8 Break-apart C-Bottom
  • Flat w/curved edge
Specifications
Specifications
Single break
Strip Plate
High Binding
96-well Microplate
96
Curved Edges, Flat Bottom (C-well)
High binding
8 x 12
White
Polystyrene
Sterile
535 x 325 x 145 mm (21.06 x 12.7 x 5.7 in.)
Product Suggestions
Product Suggestions