Learn More
Lutetium foil, 0.5mm (0.02in) thick, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 010229FF
Description
Lutetium is used as an additive for special steel, non-ferric alloy, hydrogen storage alloy, also reducer for making other rare Earth metal. Pipe materials of reactor system. Used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Lutetium foil | |
7439-94-3 | |
Lu | |
25 x 25 mm | |
Insoluble in water. | |
[Lu] | |
174.97 | |
CHEBI:33382 | |
Odorless | |
≈3.1g/25x25mm |
0.5mm (0.02 in.) thick | |
(REO) | |
MFCD00011098 | |
lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces | |
OHSVLFRHMCKCQY-UHFFFAOYSA-N | |
lutetium | |
23929 | |
99.9% | |
Mylar under Argon | |
Foil |
Chemical Identifiers
7439-94-3 | |
174.97 | |
OHSVLFRHMCKCQY-UHFFFAOYSA-N | |
23929 | |
lutetium |
Lu | |
MFCD00011098 | |
lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces | |
CHEBI:33382 | |
[Lu] |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
EINECSNumber : 231-103-0
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only