Learn More
Lutetium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
$104.97 - $862.08
Chemical Identifiers
CAS | 12032-20-1 |
---|---|
Molecular Formula | Lu2O3 |
Molecular Weight (g/mol) | 397.93 |
MDL Number | MFCD00011100 |
InChI Key | UGBIHFMRUDAMBY-UHFFFAOYSA-N |
Synonym | lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+ |
IUPAC Name | dilutetium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Lu+3].[Lu+3] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1125503
|
Thermo Scientific Chemicals
01125503 |
1 g |
Each for $104.97
|
|
|||||
AA1125506
|
Thermo Scientific Chemicals
01125506 |
5 g |
Each for $344.93
|
|
|||||
AA1125514
|
Thermo Scientific Chemicals
01125514 |
25 g |
Each for $862.08
|
|
|||||
Description
Lutetium(III) oxide serves as a catalyst in a variety of reactions such as polymerization, hydrogenation and alkylation reactions. It is also used in ceramics and glass. In the production of laser crystals, lutetium(III) oxide finds application as an important starting material. Its luminescent properties, a field of interest for researchers and may find potential application in biological fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12032-20-1 | |
397.93 | |
UGBIHFMRUDAMBY-UHFFFAOYSA-N | |
dilutetium(3+) trioxidandiide |
Lu2O3 | |
MFCD00011100 | |
lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+ | |
[O--].[O--].[O--].[Lu+3].[Lu+3] |
Specifications
2487°C | |
White | |
1 g | |
Lu2O3 | |
lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+ | |
UGBIHFMRUDAMBY-UHFFFAOYSA-N | |
dilutetium(3+) trioxidandiide | |
397.94 | |
Hygroscopic | |
9.42 |
12032-20-1 | |
Powder | |
99.9% (REO) | |
MFCD00011100 | |
Insoluble in water. | |
[O--].[O--].[O--].[Lu+3].[Lu+3] | |
397.93 | |
Odorless | |
White powder | |
Lutetium(III) oxide |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H320-H335
EINECSNumber : 234-764-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only