LW Scientific Manual Differential Counters DFS Item
Designed for blood cell counting
Supplier:  LW Scientific CTLDIFM05KY
 View more versions of this product
Catalog No. 22500100
Explore available promotions
Description
Description
  • Durable and acid-resistant
  • High-impact plastic body
  • Right and left reset knobs
  • Non-slip bottom
  • Strip chart of mature and immature forms
  • Bell alarm for 100th counted cell
  • Five and Eight key counters available
Specifications
Specifications
Five key
27.9 x 10.1 x 7.62cm (11 x 4 x 3 in.)
1.36kg
Blood cell counting