Learn More
LW Scientific Manual Differential Counters
Designed for blood cell counting
Supplier: LW Scientific CTLDIFM08KY
Description
- Durable and acid-resistant
- High-impact plastic body
- Right and left reset knobs
- Non-slip bottom
- Strip chart of mature and immature forms
- Bell alarm for 100th counted cell
- Five and Eight key counters available
Specifications
| Eight key | |
| 33 x 10.1 x 7.62cm | |
| 2.2kg | |
| Blood cell counting |