BUCHI Lyovapor™ L-200 Pro Control Freeze Dryer
The compact Lyovapor™ L-200 Pro Control offers high-quality Freeze Drying with complete automation.
Supplier:  BUCHI 11L200PH2612101102
 View more versions of this product
Catalog No. 05405438
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • A 6 L condenser capacity allows for maximum sample throughput
  • Integrating Infinite-Control™ with your freeze dryer gives you access to all process parameters and keeps you informed by push notifications anywhere, anytime
  • Fully Automated
  • Method creation and reporting
  • Real-time pressure and temperature recording
  • Stoppering available under inert atmospheric conditions
  • Small footprint
  • Touch-screen control panel
Specifications
Specifications
6 kg (Condensor)
Stainless Steel
510 mm
508.7 lb.
-55°C to +25°C
Heatable Shelves 6, Sensor, Pfeiffer Duo 6 Vacuum Pump, Mobile Trolley
208/230 V
231.3 kg