Invitrogen™ Lysing Solution, Whole Blood Lysing Solution DFS Item
Lysing Solution, Whole Blood Lysing Solution
Supplier:  Invitrogen™ GAS010
 View more versions of this product
Catalog No. 501121187
Explore available promotions
Description
Description
Invitrogen Cal-Lyse™ is a lysing solution formulated to enable the lysis of erythrocytes in samples of anticoagulated human peripheral blood. Invitrogen Cal-Lyse™ lysing solution is intended to be used as an aid in the enumeration of leukocytes that have been stained with monoclonal antibodies for flow cytometric analysis.
View a selection guide for all cell lysis and preservation solutions and products related to sample fixation and permeablization, blocking reagents, and information about the use of Dynabeads™ for cell isolation.
Specifications
Specifications
Store at room temperature.
  • Shelf life: 24 months from date of manufacture
  • Whole Blood Lysing Solution
    25 mL
    Whole Blood Lysis Buffer
    25 mL
    Room Temperature
    Flow Cytometry
    Videos
    SDS
    Documents
    Documents
    Promotions
    Promotions
    For Research Use Only. Not for use in diagnostic procedures.