missing translation for 'onlineSavingsMsg'
Learn More

m-Anisidine, 99%

CAS: 536-90-3 | C7H9NO | 123.15 g/mol

$55.02 - $187.76

Chemical Identifiers

CAS 536-90-3
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00007783
InChI Key NCBZRJODKRCREW-UHFFFAOYSA-N
Synonym m-anisidine, 3-aminoanisole, m-methoxyaniline, 3-methoxybenzenamine, 3-anisidine, m-aminoanisole, benzenamine, 3-methoxy, m-anisylamine, 1-amino-3-methoxybenzene, unii-jxa144kx2i
PubChem CID 10824
IUPAC Name 3-methoxyaniline
SMILES COC1=CC=CC(=C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC104820050
SDS View Documents
Thermo Scientific Chemicals
104820050
5 mL
Each for $55.02
Only null left
 
AC104821000
SDS View Documents
Thermo Scientific Chemicals
104821000
100 mL
Each for $187.76
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

536-90-3
123.15
NCBZRJODKRCREW-UHFFFAOYSA-N
10824
COC1=CC=CC(=C1)N
C7H9NO
MFCD00007783
m-anisidine, 3-aminoanisole, m-methoxyaniline, 3-methoxybenzenamine, 3-anisidine, m-aminoanisole, benzenamine, 3-methoxy, m-anisylamine, 1-amino-3-methoxybenzene, unii-jxa144kx2i
3-methoxyaniline

Specifications

536-90-3
1.1020g/mL
126°C
98.5% min. (GC)
C7H9NO
CH3OC6H4NH2
5 mL
1.102
m-anisidine, 3-aminoanisole, m-methoxyaniline, 3-methoxybenzenamine, 3-anisidine, m-aminoanisole, benzenamine, 3-methoxy, m-anisylamine, 1-amino-3-methoxybenzene, unii-jxa144kx2i
NCBZRJODKRCREW-UHFFFAOYSA-N
3-methoxyaniline
10824
99%
m-Anisidine, 99%
Yellow to Brown
250°C to 251°C
Authentic
Glass bottle
1.5800 to 1.582
MFCD00007783
13,404
14,667
Solubility in water: 18g/L (20°C). Other solubilities: soluble in acids,soluble in alcohol,diethyl ether,acetone and,benzene
COC1=CC=CC(=C1)N
123.15
123.15
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for

GHS Signal Word: Danger

EINECSNumber : 208-651-4

RTECSNumber : BZ5408000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only