missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Cresol Purple sodium salt, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1802514

 View more versions of this product

Catalog No. AAA1802514


Explore available promotions

Description

Description

m-Cresol purple sodium salt is used to detect changes in pH during titrations at which color changes occur at pH 7.6-9.2. Upon bromination, m-cresol purple sodium salt creates the dye bromocresol green (sc-206036) which is used as a tracking dye in DNA agarose gel electrophoresis. In addition, m-cresol purple sodium salt can be used to stain epithelial cells and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Cresol purple sodium salt is used to detect changes in pH during titrations at which color changes occur at pH 7.6-9.2. Upon bromination, m-cresol purple sodium salt creates the dye bromocresol green (sc-206036) which is used as a tracking dye in DNA agarose gel electrophoresis. In addition, m-cresol purple sodium salt can be used to stain epithelial cells and proteins.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

m-Cresol Purple sodium salt
C21H17NaO5S
m-Cresol Purple, water soluble; m-Cresolsulfonephthalein sodium salt
404.42
62625-31-4
MFCD00010177
Soluble in water.
25 g

Chemical Identifiers

62625-31-4
C21H17NaO5S
MFCD00010177
m-Cresol Purple, water soluble; m-Cresolsulfonephthalein sodium salt
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 263-656-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only