missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Phenylenediamine dihydrochloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 216650250

 View more versions of this product

Catalog No. AC216650250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

m-Phenylenediamine dihydrochloride
541-69-5
100.0
Authentic
Glass bottle
C6H4(NH2)2·2HCl
25 g
10, 7164
Solubility in water: freely soluble. Other solubilities: soluble in alcohol
C1=CC(=CC(=C1)N)N.Cl.Cl
181.07
181.07
Crystalline Powder
99%
98.5
Beige to Beige-Gray
98.5% min. (Argentometry)
C6H8N2·2ClH
MFCD00012975
13, 38
benzene-1,3-diamine dihydrochloride, m-phenylenediamine dihydrochloride, 1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine dihydrochloride, 1,3-benzenediamine, dihydrochloride, m-phenylenediamine hydrochloride, unii-se5rlo93tg, 1,3-diaminobenzene dihydrochloride, m-aminoaniline dihydrochloride, 3-aminoaniline dihydrochloride
SVTOYMIYCMHPIV-UHFFFAOYSA-N
benzene-1,3-diamine;dihydrochloride
10941
99%

Chemical Identifiers

541-69-5
181.07
SVTOYMIYCMHPIV-UHFFFAOYSA-N
10941
C1=CC(=CC(=C1)N)N.Cl.Cl
C6H8N2·2ClH
MFCD00012975
benzene-1,3-diamine dihydrochloride, m-phenylenediamine dihydrochloride, 1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine dihydrochloride, 1,3-benzenediamine, dihydrochloride, m-phenylenediamine hydrochloride, unii-se5rlo93tg, 1,3-diaminobenzene dihydrochloride, m-aminoaniline dihydrochloride, 3-aminoaniline dihydrochloride
benzene-1,3-diamine;dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-790-0

RTECSNumber : SS9800000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only