missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
m-Tolualdehyde 2,4-Dinitrophenylhydrazone 98.0+%, TCI America™
Supplier: TCI America T144810G
Specifications
m-Tolualdehyde 2,4-Dinitrophenylhydrazone | |
213°C | |
MFCD00191585 | |
1325 | |
PQBRXQUGVASQBU-UHFFFAOYSA-N | |
N-[(E)-(3-methylphenyl)methylideneamino]-2,4-dinitroaniline | |
9604108 | |
≥98.0% (HPLC,T) |
2880-05-9 | |
C14H12N4O4 | |
10 g | |
m-tolualdehyde 2,4-dinitrophenylhydrazone, m-tolualdehyde, 2,4-dinitrophenyl hydrazone, benzaldehyde, 3-methyl-, 2,4-dinitrophenyl hydrazone, 3'-methylbenzaldehyde 2,4-dinitrophenylhydrazone, m-tolualdehyd-2,4-dinitrophenylhydrazone, m-tolualdehyde 2,4-dinitro-phenylhydrazone, 3-methylbenzaldehyde 2,4-dinitrophenyl hydrazone #, 1-2,4-dinitrophenyl-2-3-methylbenzylidene hydrazine, n-e-3-methylphenyl methylideneamino-2,4-dinitroaniline, m-tolualdehyde 2,4-dinitrophenylhydrazone, environmental standard | |
CC1=CC=CC(=C1)C=NNC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-] | |
300.27 | |
300.27 | |
Crystalline Powder |
Chemical Identifiers
2880-05-9 | |
300.27 | |
PQBRXQUGVASQBU-UHFFFAOYSA-N | |
9604108 | |
CC1=CC=CC(=C1)C=NNC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-] |
C14H12N4O4 | |
MFCD00191585 | |
m-tolualdehyde 2,4-dinitrophenylhydrazone, m-tolualdehyde, 2,4-dinitrophenyl hydrazone, benzaldehyde, 3-methyl-, 2,4-dinitrophenyl hydrazone, 3'-methylbenzaldehyde 2,4-dinitrophenylhydrazone, m-tolualdehyd-2,4-dinitrophenylhydrazone, m-tolualdehyde 2,4-dinitro-phenylhydrazone, 3-methylbenzaldehyde 2,4-dinitrophenyl hydrazone #, 1-2,4-dinitrophenyl-2-3-methylbenzylidene hydrazine, n-e-3-methylphenyl methylideneamino-2,4-dinitroaniline, m-tolualdehyde 2,4-dinitrophenylhydrazone, environmental standard | |
N-[(E)-(3-methylphenyl)methylideneamino]-2,4-dinitroaniline |
Safety and Handling
TSCA : No