missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Toluic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 139050010

 View more versions of this product

Catalog No. AC139050010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

m-Toluic acid
99-04-7
100.0
Beige-Yellow to Yellow
263°C
Authentic
Plastic bottle
CH3C6H4CO2H
1 kg
14, 9535
Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol and ether
CC1=CC=CC(=C1)C(O)=O
136.15
CHEBI:10589
99%
99%
98.5
108°C to 112°C
1.05
150°C
99%
C8H8O2
MFCD00002523
09, 475
m-toluic acid, m-toluylic acid, 3-toluic acid, benzoic acid, 3-methyl, meta-toluic acid, m-methylbenzoic acid, beta-methylbenzoic acid, 3-methyl-benzoic acid, beta-bethylbenzoic acid, unii-1ua7k8eext
GPSDUZXPYCFOSQ-UHFFFAOYSA-N
3-methylbenzoic acid
7418
136.15
Crystalline Solid

Chemical Identifiers

99-04-7
136.15
GPSDUZXPYCFOSQ-UHFFFAOYSA-N
7418
3-methylbenzoic acid
C8H8O2
MFCD00002523
m-toluic acid, m-toluylic acid, 3-toluic acid, benzoic acid, 3-methyl, meta-toluic acid, m-methylbenzoic acid, beta-methylbenzoic acid, 3-methyl-benzoic acid, beta-bethylbenzoic acid, unii-1ua7k8eext
CHEBI:10589
CC1=CC=CC(=C1)C(O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-723-9

RTECSNumber : XU1200000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only