Learn More
m-Toluoyl chloride, 99%, Thermo Scientific Chemicals
$52.74 - $460.19
Chemical Identifiers
CAS | 1711-06-4 |
---|---|
Molecular Formula | C8H7ClO |
Molecular Weight (g/mol) | 154.593 |
MDL Number | MFCD00000681 |
InChI Key | YHOYYHYBFSYOSQ-UHFFFAOYSA-N |
Synonym | m-toluoyl chloride, m-methylbenzoyl chloride, 3-toluoyl chloride, toluoyl chloride, benzoyl chloride, 3-methyl, m-toluyl chloride, meta methyl benzoyl chloride, 3-methylbenzoylchloride, 3-toluoylchloride, unii-w2vjw4350k |
PubChem CID | 74375 |
IUPAC Name | 3-methylbenzoyl chloride |
SMILES | CC1=CC=CC(=C1)C(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2391414
|
Thermo Scientific Chemicals
B2391414 |
25 g |
Each for $52.74
|
|
|||||
AAB2391422
|
Thermo Scientific Chemicals
B2391422 |
100 g |
Each for $115.63
|
|
|||||
AAB2391436
|
Thermo Scientific Chemicals
B2391436 |
500 g |
Each for $460.19
|
|
|||||
Description
m-Toluoyl chloride has been used to derivatize the C1-C4 aliphatic amines for their determination in air by HPLC. It has been used in determination of atmospheric ammonia by denuder-sampling and HPLC-UV detection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsm-Toluoyl chloride has been used to derivatize the C1-C4 aliphatic amines for their determination in air by HPLC. It has been used in determination of atmospheric ammonia by denuder-sampling and HPLC-UV detection.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases, active metals, amines and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
Chemical Identifiers
1711-06-4 | |
154.593 | |
YHOYYHYBFSYOSQ-UHFFFAOYSA-N | |
74375 | |
CC1=CC=CC(=C1)C(=O)Cl |
C8H7ClO | |
MFCD00000681 | |
m-toluoyl chloride, m-methylbenzoyl chloride, 3-toluoyl chloride, toluoyl chloride, benzoyl chloride, 3-methyl, m-toluyl chloride, meta methyl benzoyl chloride, 3-methylbenzoylchloride, 3-toluoylchloride, unii-w2vjw4350k | |
3-methylbenzoyl chloride |
Specifications
1711-06-4 | |
1.173 | |
76°C (168°F) | |
C8H7ClO | |
MFCD00000681 | |
UN3265 | |
Moisture sensitive | |
Reacts with water. | |
CC1=CC=CC(=C1)C(=O)Cl | |
154.593 | |
154.6 | |
m-Toluoyl chloride |
-23°C | |
220°C | |
Pungent | |
1.5485 | |
25 g | |
878419 | |
m-toluoyl chloride, m-methylbenzoyl chloride, 3-toluoyl chloride, toluoyl chloride, benzoyl chloride, 3-methyl, m-toluyl chloride, meta methyl benzoyl chloride, 3-methylbenzoylchloride, 3-toluoylchloride, unii-w2vjw4350k | |
YHOYYHYBFSYOSQ-UHFFFAOYSA-N | |
3-methylbenzoyl chloride | |
74375 | |
99% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 216-976-8
RTECSNumber : DM6644000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only