missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Tolyl Acetate 99.0+%, TCI America™

$88.24 - $641.87

Chemical Identifiers

CAS 122-46-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00041910
InChI Key OTGAHJPFNKQGAE-UHFFFAOYSA-N
Synonym m-tolyl acetate, m-cresyl acetate, cresatin, m-acetoxytoluene, m-cresol acetate, acetic acid m-tolyl ester, cresatin-sulzberger, kresatin, acetic acid, 3-methylphenyl ester, m-methylphenyl acetate
PubChem CID 67406
IUPAC Name 3-methylphenyl acetate
SMILES CC(=O)OC1=CC=CC(C)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A065925G
View Documents
TCI America
A065925G
25 g
Each for $88.24
 
A0659500G
View Documents
TCI America
A0659500G
500 g
Each for $641.87
 
Description

Description

Specifications

Chemical Identifiers

122-46-3
150.18
OTGAHJPFNKQGAE-UHFFFAOYSA-N
67406
CC(=O)OC1=CC=CC(C)=C1
C9H10O2
MFCD00041910
m-tolyl acetate, m-cresyl acetate, cresatin, m-acetoxytoluene, m-cresol acetate, acetic acid m-tolyl ester, cresatin-sulzberger, kresatin, acetic acid, 3-methylphenyl ester, m-methylphenyl acetate
3-methylphenyl acetate

Specifications

122-46-3
Colorless
C9H10O2
25 g
OTGAHJPFNKQGAE-UHFFFAOYSA-N
3-methylphenyl acetate
67406
≥99.0% (GC)
m-Tolyl Acetate
12°C
212°C
MFCD00041910
m-tolyl acetate, m-cresyl acetate, cresatin, m-acetoxytoluene, m-cresol acetate, acetic acid m-tolyl ester, cresatin-sulzberger, kresatin, acetic acid, 3-methylphenyl ester, m-methylphenyl acetate
CC(=O)OC1=CC=CC(C)=C1
150.18
150.18
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0644

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'