missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
m-Xylene-4-sulfonic Acid Hydrate 98.0+%, TCI America™
$99.29 - $371.87
Chemical Identifiers
CAS | 88-61-9 |
---|---|
Molecular Formula | C8H10O3S |
Molecular Weight (g/mol) | 186.225 |
MDL Number | MFCD00007483 |
InChI Key | CHZLVSBMXZSPNN-UHFFFAOYSA-N |
Synonym | 2,4-Dimethylbenzenesulfonic Acid |
PubChem CID | 6938 |
IUPAC Name | 2,4-dimethylbenzenesulfonic acid |
SMILES | CC1=CC(=C(C=C1)S(=O)(=O)O)C |
Chemical Identifiers
88-61-9 | |
186.225 | |
CHZLVSBMXZSPNN-UHFFFAOYSA-N | |
6938 | |
CC1=CC(=C(C=C1)S(=O)(=O)O)C |
C8H10O3S | |
MFCD00007483 | |
2,4-Dimethylbenzenesulfonic Acid | |
2,4-dimethylbenzenesulfonic acid |
Specifications
88-61-9 | |
White | |
MFCD00007483 | |
2,4-Dimethylbenzenesulfonic Acid | |
CC1=CC(=C(C=C1)S(=O)(=O)O)C | |
186.225 | |
186.23 | |
Crystalline Powder |
61°C | |
C8H10O3S | |
25 g | |
CHZLVSBMXZSPNN-UHFFFAOYSA-N | |
2,4-dimethylbenzenesulfonic acid | |
6938 | |
≥98.0% (HPLC,T) | |
m-Xylene-4-sulfonic Acid Hydrate |
Safety and Handling
EINECSNumber : (9)-0317
RTECSNumber : ZE5075000
TSCA : Yes