missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Xylylene Diisocyanate 98.0+%, TCI America™

Supplier:  TCI America X0022100G

 View more versions of this product

Catalog No. X0022100G


Explore available promotions

Description

Description

Specifications

Specifications

m-Xylylene Diisocyanate
-7°C
126°C
MFCD00019917
2206
RTTZISZSHSCFRH-UHFFFAOYSA-N
1,3-bis(isocyanatomethyl)benzene
19262
≥98.0% (GC)
3634-83-1
Yellow
C10H8N2O2
100 g
1,3-Bis(isocyanatomethyl)benzene, Isocyanic Acid 1,3-Phenylenedimethylene Ester, m-Xylene Diisocyanate
O=C=NCC1=CC(CN=C=O)=CC=C1
188.19
188.19
Liquid

Chemical Identifiers

3634-83-1
188.19
RTTZISZSHSCFRH-UHFFFAOYSA-N
19262
O=C=NCC1=CC(CN=C=O)=CC=C1
C10H8N2O2
MFCD00019917
1,3-Bis(isocyanatomethyl)benzene, Isocyanic Acid 1,3-Phenylenedimethylene Ester, m-Xylene Diisocyanate
1,3-bis(isocyanatomethyl)benzene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2216

RTECSNumber : NR0147000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions