Leica Microsystems M80 Stereo Microscope
The Leica M80 stereo microscope offers users exceptional clarity and ease of use. Its ergonomic design ensures comfort during prolonged use. With a removable integrated camera, it allows for documentation and analysis. Elevate your inspection experience with the Leica M80 for optimal performance.
Supplier:  Leica Microsystems 8122686
Catalog No. 01-672-670
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
The Leica M80 stereo microscope is a versatile solution, excelling in inspection tasks with its ergonomic design and removable integrated camera. This microscope offers unparalleled clarity and ease of use, enhancing the efficiency of inspection processes. Its ergonomic features ensure comfortable use during prolonged periods of work. The removable integrated camera adds a layer of flexibility, allowing users to document and analyze their inspections seamlessly. Elevate your inspection experience with the Leica M80, a reliable stereo microscope that combines precision, ergonomics, and advanced imaging capabilities for optimal performance.
  • Magnification range: 7.5x -60x (1x objective, 10x eyepieces)
  • Optics parfocal
  • 8 click stop zoom settings
  • Modular system
  • Suitable for achromatic, plan or planapo objectives
  • Objective thread M60
  • ESD
Includes:
  • 10450155 Leica M80 optics carrier
  • 10450910 Eyepiece 10x/23B, adjustable, 3d gen x2
  • 10450158 Ergo Tube 10° -50° , binocular
  • 10450159 Objective achromat 1.0x, WD = 90 mm
  • 10445661 Power cable, 2m, USA
  • 10450172 Focus drive 500 mm M-Series Routine
  • 10450173 Microscope carrier
  • 10450049 Inc. light base, large w. AntiShock feet
  • 10450434 Intermediate adapter for TL and IL Base
  • 10450271 Leica LED3000 RL, 58 mm
  • 10450337 Polarization set for LED3000 RL
  • 10450266 Power Supply for LED3000/LED5000
  • 10447039 Dust cover antistatic, small
  • 12730536 Flexacam i5 (Stereo)
  • 10450859 Samsung 4k HDMI Monitor
Specifications
Specifications
100/240 V
Achromatic
Binocular
WF 10X/23 mm, Adjustable
7.5X to 60X
Educational Microscope
LED, Reflected Illumination