missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva MabSelect PrismA Chromatography Resin
Protein A Chromatography Resin
$3038.13 - $8218.46
Specifications
Matrix | Highly cross-linked agarose, spherical. |
---|---|
Content And Storage | 2 to 8°C, 20% ethanol |
Max. Operating Pressure | 300 cm/hr. |
Particle Size | 60 μm |
For Use With (Application) | Antibody affinity chromatography |
Description
MabSelect PrismA has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing future demands in mAb processing to be met.
Specifications
Highly cross-linked agarose, spherical. | |
300 cm/hr. | |
Antibody affinity chromatography | |
Alkaline-stabilized protein A-derived (E.coli). |
2 to 8°C, 20% ethanol | |
60 μm | |
Chromatography Resin |