missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mopec Maestro Encore Elevating Grossing Station

With the Maestro's distinctive Ensemble Pegboard™, dozens of accessories and flexible safety options, every station can be quickly personalized to suit each individual.

Supplier:  Mopec MB1060R

Catalog No. 22445095

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

The Maestro Encore Elevating Grossing Station provides the highest level of ergonomic support to accommodate users of varying heights, differing workspace preferences, and laboratory safety, ergonomic, and workflow requirements. Prevent workplace injuries and increase comfortability for your staff with the press of a button.

As the gold standard for safety in pathology grossing, the Mopec Maestro Encore Grossing Stations have been meticulously engineered with a focus on laboratory safety and workflow efficiency. Its patented SafeDraft™ Ventilation Technology directs air from the cutting surface’s front to the back, establishing a laminar flow pattern that enhances the effectiveness of the robust backdraft technology. This consistent ventilation is a crucial element in safeguarding users against potential hazardous formalin exposure.

The latest Mopec Maestro Encore introduces an integrated air filtration system, further optimizing airflow to mitigate and prevent formalin exposure risks. This innovative solution is designed to meet the specific demands of pathology professionals, ensuring a secure and efficient working environment.

Every Maestro Encore comes with the following standard features included:

  • TÜV Rheinland Certification
  • U.S.A. Stainless steel construction
  • Sink located on right-hand side of grossing station
  • Backdraft ventilation
  • Dual 8 in. duct stubs for in-house ventilation
  • Elevating platform for 12 in. of ergonomic height adjustment
  • OrchestraOS 2.0 Touch screen control panel and alert system
  • Patented SafeDraft™ Ventilation Technology for maximum airflow
  • Integrated Air Filtration System to prevent dust and debris build up
  • Flow Detection with backdraft alert monitoring system
  • Seismic anchoring points
  • Integrated PathCam 24.1 MP Imaging Hardware Powered by Canon
  • 60 Day PathCam Digital Pathology Free Software Trial
  • Standard water faucet with hot/cold mixing-flow control
  • Hand Spray Rinse
  • Mopec Ensemble Pegboard accessory system
  • Ergorail™ Track for mounting of monitor and keyboard arms
  • Four (4) G.F.C.I. receptacles with USB power
  • Sliding worksurface with interchangeable workflow components
  • One (1) sliding work surface perforated grid plate
  • One (1) sliding work surface dissecting board, blue
  • Two (2) wire ports for cord management
  • 1/2 Hp. disposal
  • Magnetic instrument bar
  • One (1) large and one (1) small organizer bin
Specifications

Specifications

Grossing Station
Ensemble Pegboard™ System
75 to 87 in.
32 in.
60 in.
Stainless Steel
Backdraft
60 in.
Elevating
Videos
SDS
Documents

Documents

Promotions

Promotions