missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ MagicMark™ XP Western Protein Standard
Specifically designed for easy and convenient protein molecular weight estimation directly on Western blots
Supplier: Invitrogen™ LC5602
Includes: 250 μL of ready-to-use standard mixture is provided in a plastic vial; Loading buffer consists of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue
Description
- MagicMark XP Western Protein Standard consists of nine recombinant proteins (20–220 kDa), each of which contains an IgG binding site
- The IgG binding site binds the primary or secondary antibody used for detection of the target protein, allowing direct visualization of the standard on the western blot
- The MagicMark XP Standard is compatible with most western kits and substrates (chemiluminescent, chromogenic, and fluorescent)
- The protein standard is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use
- Store at -20°C
Applications
- Western blotting: detection of the nine unstained bands via the detection method used for the target protein. Compatible with chemiluminescent substrates and fluorescent secondary antibodies (not recommended for antibodies labeled with fluors in the 500–550 nm channel)
- Protein size estimation: protein sizing on SDS-polyacrylamide gels and on western blot using western blot detection method
Specifications
250 μL of ready-to-use standard mixture is provided in a plastic vial. Loading buffer consists of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue. Store at -20°C. |
|
9 | |
Yes | |
Unstained | |
220, 120, 100, 80, 60, 50, 40, 30, 20 kDa | |
250 μL | |
Western Blotting, SDS-PAGE |
User defined detection system | |
Protein Ladder | |
20 to 220 kDa | |
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels | |
MagicMark™ | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.