Applied Biosystems™ MagMAX™ Viral/Pathogen Proteinase K
This Proteinase K is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.
Supplier: Applied Biosystems™ A42363
Description
This Proteinase K is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. No. A42352, A42356, A42357, and A42358). It is made available separately for applications that require more Proteinase K than is provided in the kit.Specifications
Store at room temperature. |
|
Next-Generation Sequencing (NGS) | |
MagMAX™ | |
10 mL | |
Viral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids |
Liquid Samples (e.g. Serum), Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast | |
High-throughput Compatible, Manual Protocols, Automated Protocols | |
Viral/Pathogen Proteinase K | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.