MilliporeSigma™ Magna ChIP™ A/G Chromatin Immunoprecipitation Kit
Analyze changes in histone modifications that correlate with processes like transcription, mitosis or DNA repair
Supplier:  MilliporeSigma™ 1710085
Catalog No. 1710085
Explore available promotions
Includes:
  • Magnetic Protein A/G Beads
  • ChIP Dilution Buffer
  • Low Salt Wash Buffer
  • High Salt Wash Buffer
  • LiCl Wash Buffer
  • TE Buffer
  • Cell Lysis Buffer
  • Nuclear Lysis Buffer
  • ChIP Elution Buffer (w/o Proteinase K)
  • Proteinase K
  • RNase A
  • 10X Glycine
  • 10X PBS
  • Protease Inhibitor Cocktail II
  • Spin Filters
  • Collection Tubes
  • Bind Reagent A
  • Wash Reagent B
  • Elution Reagent C
Requires : Magna Grip Rack 8 well
Description
Description
  • Magnetic bead-based rapid protocol allows performance of ChIP in as little as 1 day
  • Blend of protein A+G bead blend allows ChIP using a broader range of antibodies than A or G alone
  • Includes spin columns to make DNA purification easier and more reliable
  • Complete kit with all required reagents for reliable and reproducible results
  • Compatible with ChIPAb+ validated antibody and primer sets
Specifications
Specifications
Store components at the temperatures indicated on the labels
To perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads
22 Assays
Kit components are stable for 1 year from date of shipment when stored as directed