missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium chloride, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 223211000

 View more versions of this product

Catalog No. AC223211000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1412.0°C
708.0°C
White
100 g
MgCl2
11,31
magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
TWRXJAOTZQYOKJ-UHFFFAOYSA-L
magnesium;dichloride
5360315
95.21
Pure
Plastic bottle
Magnesium chloride
7786-30-3
Powder
Cl2Mg
MFCD00011106
15,5726
Solubility in water: 540g/L (20°C)
[Mg++].[Cl-].[Cl-]
95.21
CHEBI:6636
≥98.0% (on dry substance)
1.5% max. (105°C, 2 Hrs)

Chemical Identifiers

7786-30-3
95.21
TWRXJAOTZQYOKJ-UHFFFAOYSA-L
5360315
[Mg++].[Cl-].[Cl-]
Cl2Mg
MFCD00011106
magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
CHEBI:6636
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 232-094-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only