missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium EDTA, For Hardness, 0.02% (w/v), LabChem™

Supplier:  LabChem LC164501

 View more versions of this product

Catalog No. LC164501


Explore available promotions

Description

Description

Specifications

Specifications

Magnesium EDTA
For Hardness
99.98,0.02
C10H12N2O8MgNa2
1g/mL
Soluble in water
[Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
358.50
358.5
Passes Test
1g/mL
500 mL
Colorless
14402-88-1,7732-18-5
C10H12MgN2Na2O8
MFCD00078217
unii-ndt563s5vz, edta magnesium disodium, magnesium disodium edta, ndt563s5vz, magnesium disodium ethylenediaminetetraacetate, edta magnesium disodium salt hydrate, ethylenediaminetetraacetic acid magnesium disodium salt, ethylenediaminetetraacetic acid disodium magnesium salt, ethylenediaminetetraacetic acid disodium magnesium salt hydrate, magnesium sodium ethylenediaminetetraacetate
AWNVVAMWLMUZOZ-UHFFFAOYSA-J
magnesium(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
161064
0.02% (w/v)
Poly Bottle
Nitrogen oxides; Carbon monoxide; Carbon dioxide
Liquid

Chemical Identifiers

14402-88-1
358.50
AWNVVAMWLMUZOZ-UHFFFAOYSA-J
161064
[Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
C10H12MgN2Na2O8
MFCD00078217
unii-ndt563s5vz, edta magnesium disodium, magnesium disodium edta, ndt563s5vz, magnesium disodium ethylenediaminetetraacetate, edta magnesium disodium salt hydrate, ethylenediaminetetraacetic acid magnesium disodium salt, ethylenediaminetetraacetic acid disodium magnesium salt, ethylenediaminetetraacetic acid disodium magnesium salt hydrate, magnesium sodium ethylenediaminetetraacetate
magnesium(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions