Learn More
Magnesium ethoxide, 98%, Thermo Scientific Chemicals
$54.99 - $54.99
Chemical Identifiers
CAS | 2414-98-4 |
---|---|
Molecular Formula | C4H10MgO2 |
Molecular Weight (g/mol) | 114.43 |
MDL Number | MFCD00009060 |
InChI Key | XDKQUSKHRIUJEO-UHFFFAOYSA-N |
Synonym | magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide |
PubChem CID | 164963 |
IUPAC Name | magnesium;ethanolate |
SMILES | CC[O-].CC[O-].[Mg+2] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC397470050
|
Thermo Scientific Chemicals
397470050 |
5 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2414-98-4 | |
114.43 | |
XDKQUSKHRIUJEO-UHFFFAOYSA-N | |
164963 | |
CC[O-].CC[O-].[Mg+2] |
C4H10MgO2 | |
MFCD00009060 | |
magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide | |
magnesium;ethanolate |
Specifications
2414-98-4 | |
White to Gray | |
98% | |
C4H10MgO2 | |
MFCD00009060 | |
magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide | |
XDKQUSKHRIUJEO-UHFFFAOYSA-N | |
magnesium;ethanolate | |
164963 | |
98% | |
Magnesium ethoxide |
270.0°C | |
Authentic | |
Glass bottle | |
(CH3CH2O)2Mg | |
5 g | |
Solubility in water: reacts | |
CC[O-].CC[O-].[Mg+2] | |
114.43 | |
114.43 | |
Powder |
Safety and Handling
GHS H Statement
Flammable solid.
Self-heating; may catch fire.
Causes serious eye irritation.
Reacts violently with water.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
Keep cool.
Protect from sunlight.
Wash face,hands and any exposed skin thoroughly after handling.
If
GHS Signal Word: Danger
EINECSNumber : 219-323-5
RUO – Research Use Only